accessibility menu, dialog, popup

UserName

OONNHO

CAS RN 27372-38-9

IUPAC Name: 6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylate
Synonyms: 6-oxo-4,5-dihydro-1H-pyridazine-3-carboxylate
Molecular Weight (g/mol): 141.11
Molecular Formula: C5H5N2O3
InChi Key: VUADWGRLHPTYPI-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=NNC(=O)CC1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results