accessibility menu, dialog, popup

UserName

CAS RN 27304-13-8

ClOClClClClClClCl

CAS RN 27304-13-8

IUPAC Name: 1,5,6,8,9,10,11,11-octachloro-4-oxatetracyclo[6.2.1.0²,⁷.0³,⁵]undec-9-ene
Synonyms: oxychlordane
Molecular Weight (g/mol): 423.74
Molecular Formula: C10H4Cl8O
InChi Key: VWGNQYSIWFHEQU-UHFFFAOYNA-N
SMILES: ClC1C2C(C3OC13Cl)C1(Cl)C(Cl)=C(Cl)C2(Cl)C1(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results