accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 2725-82-8

IUPAC Name: 1-bromo-3-ethylbenzene
Synonyms: 1-bromo-3-ethylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: ZRFJYAZQMFCUIX-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)