accessibility menu, dialog, popup

UserName

CAS RN 27219-07-4

H3CCH3H3COOHNOOH

CAS RN 27219-07-4

IUPAC Name: 5-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 5-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: GFMRZAMDGJIWRB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results