accessibility menu, dialog, popup

UserName

H3CONHOHO

CAS RN 27115-50-0

IUPAC Name: 2-[(4-methylphenyl)formamido]acetic acid
Synonyms: [(4-methylphenyl)formamido]acetic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: NRSCPTLHWVWLLH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results