accessibility menu, dialog, popup

UserName

H3CONHN

CAS RN 27069-17-6

IUPAC Name: 5-(4-methoxyphenyl)-1H-pyrazole
Synonyms: 3-(4-methoxyphenyl)-2H-pyrazole
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: QUSABYOAMXPMQH-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=NN1
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results