accessibility menu, dialog, popup

UserName

CAS RN 27019-47-2

IUPAC Name: 3-(benzyloxy)-3-oxopropan-1-aminium
Synonyms: 3-(benzyloxy)-3-oxopropan-1-aminium
Molecular Weight (g/mol): 180.23
Molecular Formula: C10H14NO2
InChi Key: CANCPUBPPUIWPX-UHFFFAOYSA-O
SMILES: [NH3+]CCC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results