accessibility menu, dialog, popup

UserName

CAS RN 27006-82-2

IUPAC Name: 5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid
Synonyms: 5-chloro-1,3-dimethylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 174.58
Molecular Formula: C6H7ClN2O2
InChi Key: RRWQERXMLIEDKJ-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(C(O)=O)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)