accessibility menu, dialog, popup

UserName

NHN

CAS RN 27005-52-3

IUPAC Name: 2-(2-phenyl-1H-indol-3-yl)acetonitrile
Synonyms: 2-(2-phenyl-1H-indol-3-yl)acetonitrile
Molecular Weight (g/mol): 232.29
Molecular Formula: C16H12N2
InChi Key: CNAHOBNLHXZPRN-UHFFFAOYSA-N
SMILES: N#CCC1=C(NC2=CC=CC=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results