accessibility menu, dialog, popup

UserName

CAS RN 269410-08-4

CH3H3COBOH3CH3CNHN

CAS RN 269410-08-4

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Weight (g/mol): 194.04
Molecular Formula: C9H15BN2O2
InChi Key: TVOJIBGZFYMWDT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CNN=C1
Quantity 
  • (4)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)