accessibility menu, dialog, popup

UserName

H3CCH2OOOOOOCH3

CAS RN 26915-72-0

IUPAC Name: 2,5,8,11-tetraoxatridecan-13-yl 2-methylprop-2-enoate
Synonyms: 2,5,8,11-tetraoxatridecan-13-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 276.33
Molecular Formula: C13H24O6
InChi Key: KRCGBOKYIUDIFY-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOC(=O)C(C)=C
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results