CAS RN 2688-77-9
CAS RN 2688-77-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Laudanosine, TRC
CAS: 2688-77-9 Molecular Formula: C21H27NO4 Molecular Weight (g/mol): 357.44 Synonym: (1S)-1-[(3,4-Dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-isoquinoline,Laudanosine,(+)-Laudanosine,(S)-(+)-Laudanosine,(S)-Laudanosine,L-(+)-Laudanosine,L-Laudanosine,(+)-Laudanosine,,NSC 35045,O-Methylcodamine; SMILES: CN1CCC2=CC(=C(C=C2[C@@H]1CC3=CC(=C(C=C3)OC)OC)OC)OC