accessibility menu, dialog, popup

UserName

CAS RN 2672-58-4

CH3OOOOH3COOCH3

CAS RN 2672-58-4

IUPAC Name: 1,3,5-trimethyl benzene-1,3,5-tricarboxylate
Synonyms: 1,3,5-trimethyl benzene-1,3,5-tricarboxylate
Molecular Weight (g/mol): 252.22
Molecular Formula: C12H12O6
InChi Key: RGCHNYAILFZUPL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)