accessibility menu, dialog, popup

UserName

NaNNN

CAS RN 26628-22-8

IUPAC Name: sodium azide
Synonyms: sodium azide natriumazid hydrazoic acid, sodium salt azoture de sodium
Molecular Weight (g/mol): 65.01
Molecular Formula: N3Na
InChi Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES: [Na+].[N-]=[N+]=[N-]
Molecular Weight (g/mol) 
  • (12)
  • (17)
Quantity 
  • (2)
  • (3)
  • (8)
  • (1)
  • (3)
  • (1)
  • (2)
  • (9)
Physical Form 
  • (4)
  • (8)
  • (3)
  • (4)
  • (4)
Grade 
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (7)
Percent Purity 
  • (6)
  • (1)
  • (1)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (7)
  • (3)
  • (3)
  • (12)
  • (17)
  • (2)
  • (3)
  • (8)
  • (1)
  • (3)
  • (1)
  • (2)
  • (9)
  • (4)
  • (8)
  • (3)
  • (4)
  • (4)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (7)
  • (6)
  • (1)
  • (1)
  • (8)