CAS RN 26627-77-0
CAS RN 26627-77-0
IUPAC Name:
heptyl(octyl)amine
Synonyms:
heptyl(octyl)amine
Molecular Weight (g/mol):
227.44
Molecular Formula:
C15H33N
InChi Key:
HAEBNPLWJOGFDQ-UHFFFAOYSA-N
SMILES:
CCCCCCCCNCCCCCCC
1
–
1
of
1
results
1
N-(n-Heptyl)-n-octylamine, 98+%, Thermo Scientific™
CAS: 26627-77-0 Molecular Formula: C15H33N Molecular Weight (g/mol): 227.436 MDL Number: MFCD03093076 InChI Key: HAEBNPLWJOGFDQ-UHFFFAOYSA-N Synonym: n-n-heptyl-n-octylamine,heptyloctylamine,n-heptyloctylamine,heptyl-octyl-amine,n-heptyl-n-octylamine,heptyl octyl amine,1-octanamine, n-heptyl PubChem CID: 12905714 IUPAC Name: N-heptyloctan-1-amine SMILES: CCCCCCCCNCCCCCCC