accessibility menu, dialog, popup

UserName

H3CNCH3OSCl

CAS RN 26615-21-4

IUPAC Name: [2-({6-chloro-2-thiatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-9-yl}oxy)ethyl]dimethylamine
Synonyms: compound-4 lodopin nipolept zotepina zotepine aventis brand of zotepine C18H18ClNOS setous zoleptil zotepine (substance)
Molecular Weight (g/mol): 331.86
Molecular Formula: C18H18ClNOS
InChi Key: HDOZVRUNCMBHFH-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC2=CC=CC=C2SC2=CC=C(Cl)C=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results