accessibility menu, dialog, popup

UserName

CH3NH3CNCH3H3CONOOFPFFFFF

CAS RN 265651-18-1

IUPAC Name: [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
Synonyms: [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium hexafluorophosphate
Molecular Weight (g/mol): 359.21
Molecular Formula: C9H16F6N3O3P
InChi Key: STWZCCVNXFLDDD-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)