accessibility menu, dialog, popup

UserName

CAS RN 26536-60-7

H3CNHSNHCH3

CAS RN 26536-60-7

IUPAC Name: 1,3-dipropylthiourea
Synonyms: 1,3-dipropylthiourea
Molecular Weight (g/mol): 160.28
Molecular Formula: C7H16N2S
InChi Key: AUXGIIVHLRLBSG-UHFFFAOYSA-N
SMILES: CCCNC(=S)NCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results