accessibility menu, dialog, popup

UserName

CAS RN 26494-55-3

IUPAC Name: 2-(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)acetate
Synonyms: 2-(3-oxo-2H-1,4-benzoxazin-4-yl)acetate
Molecular Weight (g/mol): 206.18
Molecular Formula: C10H8NO4
InChi Key: POGLODLVBYOXAO-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1C(=O)COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results