accessibility menu, dialog, popup

UserName

CAS RN 2646-91-5

FOF

CAS RN 2646-91-5

IUPAC Name: 2,3-difluorobenzaldehyde
Synonyms: 2,3-difluorobenzaldehyde
Molecular Weight (g/mol): 142.11
Molecular Formula: C7H4F2O
InChi Key: WDBAXYQUOZDFOJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C=O)=C1F
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)