accessibility menu, dialog, popup

UserName

CAS RN 2646-90-4

FFO

CAS RN 2646-90-4

IUPAC Name: 2,5-difluorobenzaldehyde
Synonyms: 2,5-difluorobenzaldehyde
Molecular Weight (g/mol): 142.11
Molecular Formula: C7H4F2O
InChi Key: VVVOJODFBWBNBI-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(C=O)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)