CAS RN 2646-38-0
IUPAC Name:
sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate
Synonyms:
sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate
Molecular Weight (g/mol):
414.56
Molecular Formula:
C24H39NaO4
InChi Key:
WDFRNBJHDMUMBL-UHFFFAOYNA-M
SMILES:
[Na+].CC(CCC([O-])=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C
1
–
1
of
1
results
1
Chenodeoxycholic Acid Sodium Salt, TRC
CAS: 2646-38-0 Molecular Formula: C24 H39 O4 . Na Molecular Weight (g/mol): 414.554 Synonym: Chenodeoxycholic Acid Sodium Salt,Chenodesoxycholic Acid Sodium Salt,NSC 681066,Sodium Chenodeoxycholate,Sodium Chenodesoxycholate,3alpha,7alpha-Dihydroxy-5beta-cholanic Acid Sodium Salt,3alpha,7alpha-Dihydroxy-5beta-cholan-24-oic Acid Monosodium Salt IUPAC Name: sodium;(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate SMILES: [Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C