accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3C(E/Z)

CAS RN 264144-59-4

IUPAC Name: 2-(1,2-diphenylethenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(1,2-diphenylethenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 306.21
Molecular Formula: C20H23BO2
InChi Key: HGZKORGQZNCVQC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C(=CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results