accessibility menu, dialog, popup

UserName

CH3NNNH2

CAS RN 263276-44-4

IUPAC Name: 4-(4-methylphenyl)pyrimidin-2-amine
Synonyms: 4-(4-methylphenyl)pyrimidin-2-amine
Molecular Weight (g/mol): 185.23
Molecular Formula: C11H11N3
InChi Key: SEAVIOKUWQLSPU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NC(N)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results