accessibility menu, dialog, popup

UserName

CAS RN 26239-55-4

H2NONHOOOO

CAS RN 26239-55-4

IUPAC Name: 2-[(carbamoylmethyl)(carboxylatomethyl)azaniumyl]acetate
Synonyms: 2-[(carbamoylmethyl)(carboxylatomethyl)ammonio]acetate
Molecular Weight (g/mol): 189.15
Molecular Formula: C6H9N2O5
InChi Key: QZTKDVCDBIDYMD-UHFFFAOYSA-M
SMILES: NC(=O)C[NH+](CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)