CAS RN 2622-30-2
CAS RN 2622-30-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carphenazine, TRC
CAS: 2622-30-2 Molecular Formula: C24 H31 N3 O2 S Molecular Weight (g/mol): 425.59 Synonym: 1-[10-[3-[4-(2-Hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-1-propanone Carfenazine,1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]- (6CI,7CI,8CI),Carfenazine,Carphenazin,Carphenazine,Proketazine IUPAC Name: 1-[10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]propan-1-one SMILES: CCC(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(CCO)CC4)c2c1