accessibility menu, dialog, popup

UserName

CAS RN 261952-17-4

CH3FFFBr

CAS RN 261952-17-4

IUPAC Name: 2-(bromomethyl)-1-methyl-4-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-1-methyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 253.06
Molecular Formula: C9H8BrF3
InChi Key: ZQYDOVMTWZCBIH-UHFFFAOYSA-N
SMILES: CC1=C(CBr)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results