accessibility menu, dialog, popup

UserName

CAS RN 261952-16-3

CH3BrFFF

CAS RN 261952-16-3

IUPAC Name: 1-(bromomethyl)-2-methyl-3-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-methyl-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 253.06
Molecular Formula: C9H8BrF3
InChi Key: YSABBOPLIUMXKY-UHFFFAOYSA-N
SMILES: CC1=C(CBr)C=CC=C1C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results