accessibility menu, dialog, popup

UserName

CH3OFFFN

CAS RN 261951-87-5

IUPAC Name: 4-methoxy-3-(trifluoromethyl)benzonitrile
Synonyms: 4-methoxy-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: FLHQJZPLFRIGOT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C(F)(F)F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results