accessibility menu, dialog, popup

UserName

CAS RN 26177-44-6

HClH2NBr

CAS RN 26177-44-6

IUPAC Name: hydrogen 1-(4-bromophenyl)methanamine chloride
Synonyms: hydrogen 4-bromo-benzylamine chloride
Molecular Weight (g/mol): 222.51
Molecular Formula: C7H9BrClN
InChi Key: BVYUYDBWQMSOKM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results