accessibility menu, dialog, popup

UserName

CH3BrFF

CAS RN 261763-43-3

IUPAC Name: 1-(bromomethyl)-2,3-difluoro-4-methylbenzene
Synonyms: 1-(bromomethyl)-2,3-difluoro-4-methylbenzene
Molecular Weight (g/mol): 221.05
Molecular Formula: C8H7BrF2
InChi Key: YAPOYZMZHWXAOX-UHFFFAOYSA-N
SMILES: CC1=CC=C(CBr)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results