accessibility menu, dialog, popup

UserName

FFFClN

CAS RN 261763-26-2

IUPAC Name: 2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 219.59
Molecular Formula: C9H5ClF3N
InChi Key: XCPSZOAJWIBKNE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results