accessibility menu, dialog, popup

UserName

CAS RN 261762-79-2

CH3FOCH3Cl

CAS RN 261762-79-2

IUPAC Name: 2-chloro-4-fluoro-3-methoxy-1-methylbenzene
Synonyms: 2-chloro-4-fluoro-3-methoxy-1-methylbenzene
Molecular Weight (g/mol): 174.6
Molecular Formula: C8H8ClFO
InChi Key: YFEYEJLNBZOHSD-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(C)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results