accessibility menu, dialog, popup

UserName

CAS RN 26137-08-6

CH3OOSBr

CAS RN 26137-08-6

IUPAC Name: methyl 3-bromothiophene-2-carboxylate
Synonyms: methyl 3-bromothiophene-2-carboxylate
Molecular Weight (g/mol): 221.07
Molecular Formula: C6H5BrO2S
InChi Key: PEGSJNCGPSIJOX-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Br)C=CS1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)