accessibility menu, dialog, popup

UserName

CAS RN 2612-57-9

ClNCOCl

CAS RN 2612-57-9

IUPAC Name: 2,4-dichloro-1-isocyanatobenzene
Synonyms: 2,4-dichloro-1-isocyanatobenzene
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: OLBJNSPBWLCTOT-UHFFFAOYSA-N
SMILES: ClC1=CC=C(N=C=O)C(Cl)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results