accessibility menu, dialog, popup

UserName

AgFPFFFFF

CAS RN 26042-63-7

IUPAC Name: silver(1+) hexafluoro-λ⁵-phosphanuide
Synonyms: silver hexafluorophosphate
Molecular Weight (g/mol): 252.83
Molecular Formula: AgF6P
InChi Key: SCQBROMTFBBDHF-UHFFFAOYSA-N
SMILES: [Ag+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results