CAS RN 25987-06-8
CAS RN 25987-06-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Polyethylenimine (Branched) (Technical Grade), TRC
CAS: 25987-06-8 Synonym: Ethylenediamine-ethylenimine copolymer,Aziridine-1,2-diaminoethane copolymer,Ethylenediamine, polymer with ethylenimine IUPAC Name: N'-[2-[2-[2-(2-aminoethylamino)ethyl-[2-[bis(2-aminoethyl)amino]ethyl]amino]ethyl-[2-[2-[bis(2-aminoethyl)amino]ethylamino]ethyl]amino]ethyl]ethane-1,2-diamine SMILES: NCCNCCN(CCNCCN(CCN)CCN)CCN(CCNCCN)CCN(CCN)CCN