accessibility menu, dialog, popup

UserName

NN(?)HClNCH3MeMe

CAS RN 25952-53-8

IUPAC Name: {3-[cyano(ethyl)amino]propyl}dimethylazanium chloride
Synonyms: {3-[cyano(ethyl)amino]propyl}dimethylazanium chloride
Molecular Weight (g/mol): 191.7
Molecular Formula: C8H18ClN3
InChi Key: FDXPUDRRFDHONO-UHFFFAOYSA-N
SMILES: [Cl-].CCN(CCC[NH+](C)C)C#N
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)