accessibility menu, dialog, popup

UserName

NNNN

CAS RN 25939-87-1

IUPAC Name: pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Synonyms: pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H4N4
InChi Key: RRHORVAOECWFPT-UHFFFAOYSA-N
SMILES: N#CC1=C2N=CC=CN2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results