accessibility menu, dialog, popup

UserName

FFFOHOClCl

CAS RN 25922-43-4

IUPAC Name: 2,3-dichloro-6-(trifluoromethyl)benzoic acid
Synonyms: 2,3-dichloro-6-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 259.01
Molecular Formula: C8H3Cl2F3O2
InChi Key: BHQKUOSDTCYXEE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC(Cl)=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results