accessibility menu, dialog, popup

UserName

CAS RN 25922-41-2

FFFClOHOCl

CAS RN 25922-41-2

IUPAC Name: 2,6-dichloro-3-(trifluoromethyl)benzoic acid
Synonyms: 2,6-dichloro-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 259.01
Molecular Formula: C8H3Cl2F3O2
InChi Key: WRNSEKNHEIXBES-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC(=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results