accessibility menu, dialog, popup

UserName

BFOHHOOH

CAS RN 259209-20-6

IUPAC Name: (5-fluoro-2-hydroxyphenyl)boronic acid
Synonyms: 5-fluoro-2-hydroxyphenylboronic acid
Molecular Weight (g/mol): 155.92
Molecular Formula: C6H6BFO3
InChi Key: TWDQSJDFXUMAOI-UHFFFAOYSA-N
SMILES: OB(O)C1=C(O)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results