accessibility menu, dialog, popup

UserName

OOO

CAS RN 25871-69-6

IUPAC Name: (3,5-dibenzoylphenyl)(phenyl)methanone
Synonyms: (3,5-dibenzoylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 390.44
Molecular Formula: C27H18O3
InChi Key: FDSRSUAVHPFWGT-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC(=CC(=C1)C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results