accessibility menu, dialog, popup

UserName

CAS RN 25699-82-5

IUPAC Name: 3-(1H-pyrazol-1-yl)benzonitrile
Synonyms: 3-(pyrazol-1-yl)benzonitrile
Molecular Weight (g/mol): 169.19
Molecular Formula: C10H7N3
InChi Key: KLZKYUMVLIQOFX-UHFFFAOYSA-N
SMILES: N#CC1=CC(=CC=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results