accessibility menu, dialog, popup

UserName

CAS RN 2567-29-5

Br

CAS RN 2567-29-5

IUPAC Name: 4-(bromomethyl)-1,1'-biphenyl
Synonyms: 4-(bromomethyl)-1,1'-biphenyl
Molecular Weight (g/mol): 247.14
Molecular Formula: C13H11Br
InChi Key: HZQLUIZFUXNFHK-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results