accessibility menu, dialog, popup

UserName

CAS RN 2565-58-4

OHPOHOCl

CAS RN 2565-58-4

IUPAC Name: (chloromethyl)phosphonic acid
Synonyms: chloromethylphosphonic acid
Molecular Weight (g/mol): 130.46
Molecular Formula: CH4ClO3P
InChi Key: MOFCYHDQWIZKMY-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCl

 No Refinements