accessibility menu, dialog, popup

UserName

NO

CAS RN 25562-98-5

IUPAC Name: 2-(2-phenoxyphenyl)acetonitrile
Synonyms: 2-(2-phenoxyphenyl)acetonitrile
Molecular Weight (g/mol): 209.25
Molecular Formula: C14H11NO
InChi Key: PCMQCXFXODBYGN-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CC=C1OC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results