CAS RN 25503-91-7
CAS RN 25503-91-7
IUPAC Name:
1-acetylpiperidine-4-carbonitrile
Synonyms:
1-acetylpiperidine-4-carbonitrile
Molecular Weight (g/mol):
152.2
Molecular Formula:
C8H12N2O
InChi Key:
NFDGRMQIOHRQHF-UHFFFAOYSA-N
SMILES:
CC(=O)N1CCC(CC1)C#N
1
–
1
of
1
results
1
1-Acetylpiperidine-4-carbonitrile, 97%, Thermo Scientific™
CAS: 25503-91-7 Molecular Formula: C8H12N2O Molecular Weight (g/mol): 152.20 MDL Number: MFCD00221055 InChI Key: NFDGRMQIOHRQHF-UHFFFAOYSA-N Synonym: 1-acetyl-4-piperidinecarbonitrile,1-acetyl-4-cyanopiperidine,1-acetyl-isonipecotonitrile,4-piperidinecarbonitrile,1-acetyl,1-ethanoylpiperidine-4-carbonitrile PubChem CID: 2756286 IUPAC Name: 1-acetylpiperidine-4-carbonitrile SMILES: CC(=O)N1CCC(CC1)C#N