accessibility menu, dialog, popup

UserName

CAS RN 25496-72-4

H3COOOHOH(Z)

CAS RN 25496-72-4

IUPAC Name: 2,3-dihydroxypropyl (9Z)-octadec-9-enoate
Synonyms: oleoyl glycerol
Molecular Weight (g/mol): 356.55
Molecular Formula: C21H40O4
InChi Key: RZRNAYUHWVFMIP-KTKRTIGZNA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results