accessibility menu, dialog, popup

UserName

CAS RN 25440-14-6

HNNNNHFFFFFFFFFFFFFFFFFFFF

CAS RN 25440-14-6

IUPAC Name: 2,7,12,17-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 2,7,12,17-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 974.56
Molecular Formula: C44H10F20N4
InChi Key: GBAPBSUXEFFUAD-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(F)=C1F)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results